Assay Details
| Gene Symbol: GART-B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015300095.2 | XP_015155581.1 | 20 - 21 | 3150 | 123 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GART-B| Gene Symbol: GART-B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015300095.2 | XP_015155581.1 | 20 - 21 | 3150 | 123 | |
Set Membership: |
|