Assay Details
| Gene Symbol: LOC101748706 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025148127.1 | XP_025003895.1 | 9 - 10 | 3012 | 124 | |
| XM_025148128.1 | XP_025003896.1 | 10 - 11 | 3290 | 124 | ||
| XM_025148129.1 | XP_025003897.1 | 9 - 10 | 3007 | 124 | ||
| XM_025148130.1 | XP_025003898.1 | 10 - 11 | 2562 | 124 | ||
| XM_025148131.1 | XP_025003899.1 | 9 - 10 | 3012 | 124 | ||
| XM_025148132.1 | XP_025003900.1 | 9 - 10 | 3012 | 124 | ||
| XM_025148133.1 | XP_025003901.1 | 9 - 10 | 3012 | 124 | ||
| XM_025148134.1 | XP_025003902.1 | 9 - 10 | 3012 | 124 | ||
More Information
Set Membership: |
|

LOC101748706
Genomic Map