Assay Details
| Gene Symbol: NCKAP5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025145399.1 | XP_025001167.1 | 11 - 12 | 2648 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NCKAP5L| Gene Symbol: NCKAP5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025145399.1 | XP_025001167.1 | 11 - 12 | 2648 | 85 | |
Set Membership: |
|