Assay Details
| Gene Symbol: SP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015300329.2 | XP_015155815.2 | 1 - 2 | 253 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015300329.2 transcripts.
Set Membership: |
|

SP7
Genomic Map