Assay Details
| Gene Symbol: LOC107049790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025145718.1 | XP_025001486.1 | 21 - 22 | 3511 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC107049790| Gene Symbol: LOC107049790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025145718.1 | XP_025001486.1 | 21 - 22 | 3511 | 70 | |
Set Membership: |
|