Assay Details
| Gene Symbol: MACC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015281864.2 | XP_015137350.1 | 1 - 2 | 1619 | 94 | |
| XM_418705.6 | XP_418705.4 | 1 - 2 | 117 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015281864.2, XM_418705.6 transcripts.
Set Membership: |
|

MACC1
Genomic Map