Assay Details
Gene Symbol: OTOP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015295461.2 | XP_015150947.1 | 3 - 4 | 684 | 55 | |
XM_420115.5 | XP_420115.3 | 3 - 4 | 684 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
