Assay Details
| Gene Symbol: NEUROD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003643420.4 | XP_003643468.1 | 1 - 2 | 188 | 85 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003643420.4 transcripts.
Set Membership: |
|

NEUROD2
Genomic Map