Assay Details
| Gene Symbol: ISX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025155528.1 | XP_025011296.1 | 3 - 4 | 975 | 83 | |
| XM_025155533.1 | XP_025011301.1 | 2 - 3 | 833 | 83 | ||
| XM_025155535.1 | XP_025011303.1 | 2 - 3 | 990 | 83 | ||
| XM_416296.6 | XP_416296.2 | 3 - 4 | 622 | 83 | ||
More Information
Set Membership: |
|

ISX
Genomic Map