Assay Details
| Gene Symbol: ARSE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004938437.3 | XP_004938494.1 | 10 - 11 | 1581 | 74 | |
| XM_004938438.3 | XP_004938495.1 | 10 - 11 | 1537 | 74 | ||
| XM_416856.6 | XP_416856.4 | 10 - 11 | 1591 | 74 | ||
More Information
Set Membership: |
|

ARSE
Genomic Map