Assay Details
Gene Symbol: TMPRSS11F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004941191.3 | XP_004941248.1 | 7 - 8 | 3360 | 101 | |
XM_025150261.1 | XP_025006029.1 | 7 - 8 | 3366 | 101 | ||
XM_025150262.1 | XP_025006030.1 | 7 - 8 | 3331 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
