Assay Details
| Gene Symbol: TMPRSS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025143307.1 | XP_024999075.1 | 9 - 10 | 1731 | 63 | |
| XM_025143308.1 | XP_024999076.1 | 10 - 11 | 1647 | 63 | ||
| XM_025143309.1 | XP_024999077.1 | 10 - 11 | 1638 | 63 | ||
| XM_025143310.1 | XP_024999078.1 | 9 - 10 | 1536 | 63 | ||
| XM_025143311.1 | XP_024999079.1 | 10 - 11 | 1648 | 63 | ||
| XM_025143312.1 | XP_024999080.1 | 11 - 12 | 1555 | 63 | ||
More Information
Set Membership: |
|

TMPRSS5
Genomic Map