Assay Details
| Gene Symbol: PLA2G4EL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001234815.5 | XP_001234816.3 | 5 - 6 | 553 | 129 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLA2G4EL5| Gene Symbol: PLA2G4EL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001234815.5 | XP_001234816.3 | 5 - 6 | 553 | 129 | |
Set Membership: |
|