Assay Details
| Gene Symbol: LOC101748436 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004939058.3 | XP_004939115.2 | 1 - 2 | 294 | 77 | |
| XM_025147387.1 | XP_025003155.1 | 1 - 2 | 295 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004939058.3, XM_025147387.1 transcripts.
Set Membership: |
|

LOC101748436
Genomic Map