Assay Details
| Gene Symbol: LOC101748436 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004939058.3 | XP_004939115.2 | 7 - 8 | 1181 | 108 | |
| XM_025147387.1 | XP_025003155.1 | 7 - 8 | 1149 | 108 | ||
More Information
Set Membership: |
|

LOC101748436
Genomic Map