Assay Details
| Gene Symbol: CDK2AP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015286303.2 | XP_015141789.1 | 2 - 3 | 240 | 84 | |
| XM_015286304.2 | XP_015141790.1 | 2 - 3 | 306 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015286304.2 transcripts.
Set Membership: |
|

CDK2AP2
Genomic Map