Assay Details
| Gene Symbol: LOC107055388 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025144459.1 | XP_025000227.1 | 14 - 15 | 1120 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC107055388| Gene Symbol: LOC107055388 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025144459.1 | XP_025000227.1 | 14 - 15 | 1120 | 96 | |
Set Membership: |
|