Assay Details
| Gene Symbol: XKR7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001235082.4 | XP_001235083.3 | 2 - 3 | 712 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

XKR7| Gene Symbol: XKR7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001235082.4 | XP_001235083.3 | 2 - 3 | 712 | 77 | |
Set Membership: |
|