Assay Details
| Gene Symbol: SYT10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004937879.3 | XP_004937936.1 | 2 - 3 | 665 | 70 | |
| XM_416361.6 | XP_416361.2 | 2 - 3 | 665 | 70 | ||
More Information
Set Membership: |
|

SYT10
Genomic Map