Assay Details
| Gene Symbol: PLA2G4EL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015287109.2 | XP_015142595.1 | 13 - 14 | 1319 | 94 | |
| XM_015287110.2 | XP_015142596.1 | 13 - 14 | 1313 | 94 | ||
| XM_015287112.2 | XP_015142598.1 | 13 - 14 | 1466 | 94 | ||
| XM_025150925.1 | XP_025006693.1 | 11 - 12 | 1234 | 94 | ||
| XM_025150926.1 | XP_025006694.1 | 13 - 14 | 4000 | 94 | ||
More Information
Set Membership: |
|

PLA2G4EL1
Genomic Map