Assay Details
| Gene Symbol: PLA2G4EL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015287109.2 | XP_015142595.1 | 1 - 2 | 173 | 95 | |
| XM_015287110.2 | XP_015142596.1 | 1 - 2 | 170 | 95 | ||
| XM_015287112.2 | XP_015142598.1 | 1 - 2 | 320 | 95 | ||
| XM_025150925.1 | XP_025006693.1 | 1 - 2 | 172 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015287109.2, XM_015287110.2, XM_015287112.2, XM_025150925.1 transcripts.
Set Membership: |
|

PLA2G4EL1
Genomic Map