Assay Details
| Gene Symbol: MUC4L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025153623.1 | XP_025009391.1 | 9 - 10 | 3513 | 88 | |
| XM_025153624.1 | XP_025009392.1 | 8 - 9 | 3465 | 88 | ||
More Information
Set Membership: |
|

MUC4L
Genomic Map