Assay Details
| Gene Symbol: MUC4L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025153623.1 | XP_025009391.1 | 5 - 6 | 2963 | 86 | |
| XM_025153624.1 | XP_025009392.1 | 4 - 5 | 2915 | 86 | ||
More Information
Set Membership: |
|

MUC4L
Genomic Map