Assay Details
| Gene Symbol: LOC427816 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015295890.2 | XP_015151376.1 | 2 - 3 | 244 | 200 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC427816| Gene Symbol: LOC427816 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015295890.2 | XP_015151376.1 | 2 - 3 | 244 | 200 | |
Set Membership: |
|