Assay Details
| Gene Symbol: LOC101748851 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025142945.1 | XP_024998713.1 | 5 - 6 | 772 | 93 | |
| XM_025142946.1 | XP_024998714.1 | 5 - 6 | 772 | 93 | ||
| XM_025142947.1 | XP_024998715.1 | 5 - 6 | 772 | 93 | ||
| XM_025142948.1 | XP_024998716.1 | 5 - 6 | 772 | 93 | ||
| XM_025142949.1 | XP_024998717.1 | 5 - 6 | 772 | 93 | ||
More Information
Set Membership: |
|

LOC101748851
Genomic Map