Assay Details
Gene Symbol: LOC101749673 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025145791.1 | XP_025001559.1 | 1 - 2 | 223 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025145791.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
