Assay Details
| Gene Symbol: LOC101749673 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025145791.1 | XP_025001559.1 | 2 - 3 | 273 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025145791.1 transcripts.
Set Membership: |
|

LOC101749673
Genomic Map