Assay Details
| Gene Symbol: TMEM170B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015275982.2 | XP_015131468.1 | 1 - 2 | 658 | 53 | |
| XM_418945.6 | XP_418945.1 | 1 - 2 | 510 | 53 | ||
More Information
Set Membership: |
|

TMEM170B
Genomic Map