Assay Details
| Gene Symbol: LOC107052489 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015281173.2 | XP_015136659.1 | 6 - 7 | 5941 | 72 | |
| XM_025147380.1 | XP_025003148.1 | 7 - 8 | 4999 | 72 | ||
| XM_025147381.1 | XP_025003149.1 | 7 - 8 | 4978 | 72 | ||
| XM_025147382.1 | XP_025003150.1 | 7 - 8 | 4119 | 72 | ||
More Information
Set Membership: |
|

LOC107052489
Genomic Map