Assay Details
| Gene Symbol: OCSTAMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015296720.2 | XP_015152206.1 | 1 - 2 | 2341 | 160 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015296720.2 transcripts.
Set Membership: |
|

OCSTAMP
Genomic Map