Assay Details
| Gene Symbol: LOC425940 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025142802.1 | XP_024998570.1 | 6 - 7 | 722 | 127 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC425940| Gene Symbol: LOC425940 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025142802.1 | XP_024998570.1 | 6 - 7 | 722 | 127 | |
Set Membership: |
|