Assay Details
| Gene Symbol: SEPT12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004945415.3 | XP_004945472.1 | 5 - 6 | 1420 | 96 | |
| XM_004945416.2 | XP_004945473.1 | 4 - 5 | 683 | 96 | ||
| XM_004945417.3 | XP_004945474.1 | 4 - 5 | 1133 | 96 | ||
| XM_015294708.2 | XP_015150194.1 | 4 - 5 | 567 | 96 | ||
| XM_015294710.2 | XP_015150196.1 | 4 - 5 | 712 | 96 | ||
| XM_015294711.2 | XP_015150197.1 | 4 - 5 | 519 | 96 | ||
More Information
Set Membership: |
|

SEPT12
Genomic Map