Assay Details
| Gene Symbol: CDC20B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025144998.1 | XP_025000766.1 | 4 - 5 | 576 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDC20B| Gene Symbol: CDC20B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025144998.1 | XP_025000766.1 | 4 - 5 | 576 | 86 | |
Set Membership: |
|