Assay Details
| Gene Symbol: FAM194A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015291842.2 | XP_015147328.1 | 2 - 3 | 2986 | 84 | |
| XM_015291843.2 | XP_015147329.1 | 3 - 4 | 2896 | 84 | ||
More Information
Set Membership: |
|

FAM194A
Genomic Map