Assay Details
Gene Symbol: TRIM50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015295904.2 | XP_015151390.1 | 3 - 4 | 829 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
