Assay Details
| Gene Symbol: STX11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004935577.3 | XP_004935634.1 | 2 - 3 | 326 | 68 | |
| XM_015284312.2 | XP_015139798.1 | 2 - 3 | 590 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004935577.3, XM_015284312.2 transcripts.
Set Membership: |
|

STX11
Genomic Map