Assay Details
| Gene Symbol: NR4A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015273518.2 | XP_015129004.2 | 6 - 7 | 1553 | 51 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NR4A1| Gene Symbol: NR4A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015273518.2 | XP_015129004.2 | 6 - 7 | 1553 | 51 | |
Set Membership: |
|