Assay Details
| Gene Symbol: LIPML3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015278695.2 | XP_015134181.1 | 6 - 7 | 6254 | 74 | |
| XM_015278696.2 | XP_015134182.1 | 7 - 8 | 4694 | 74 | ||
| XM_015278697.2 | XP_015134183.1 | 7 - 8 | 5407 | 74 | ||
| XM_015278699.2 | XP_015134185.1 | 7 - 8 | 5208 | 74 | ||
More Information
Set Membership: |
|

LIPML3
Genomic Map