Assay Details
| Gene Symbol: C2H8ORF34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025147990.1 | XP_025003758.1 | 4 - 5 | 870 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C2H8ORF34| Gene Symbol: C2H8ORF34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025147990.1 | XP_025003758.1 | 4 - 5 | 870 | 78 | |
Set Membership: |
|