Assay Details
| Gene Symbol: MXD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015293860.2 | XP_015149346.1 | 3 - 4 | 744 | 62 | |
| XM_015293861.2 | XP_015149347.1 | 2 - 3 | 714 | 62 | ||
| XM_015293862.2 | XP_015149348.1 | 3 - 4 | 693 | 62 | ||
More Information
Set Membership: |
|

MXD3
Genomic Map