Assay Details
| Gene Symbol: ELOF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015302698.2 | XP_015158184.1 | 2 - 3 | 461 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ELOF1| Gene Symbol: ELOF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015302698.2 | XP_015158184.1 | 2 - 3 | 461 | 75 | |
Set Membership: |
|