Assay Details
| Gene Symbol: POU3F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015284671.2 | XP_015140157.1 | 1 | 167 | 111 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015284671.2 transcripts.
Set Membership: |
|

POU3F2
Genomic Map