Assay Details
| Gene Symbol: HPCA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001233528.5 | XP_001233529.3 | 2 - 3 | 559 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HPCA| Gene Symbol: HPCA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001233528.5 | XP_001233529.3 | 2 - 3 | 559 | 67 | |
Set Membership: |
|