Assay Details
| Gene Symbol: ATXN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003640755.4 | XP_003640803.1 | 1 - 2 | 441 | 107 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003640755.4 transcripts.
Set Membership: |
|

ATXN1
Genomic Map