Assay Details
| Gene Symbol: OTOP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003642368.4 | XP_003642416.1 | 6 - 7 | 1192 | 64 | |
| XM_004946360.3 | XP_004946417.1 | 6 - 7 | 1196 | 64 | ||
| XM_015295462.2 | XP_015150948.1 | 6 - 7 | 1304 | 64 | ||
More Information
Set Membership: |
|

OTOP2
Genomic Map