Assay Details
| Gene Symbol: EFCAB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004948522.3 | XP_004948579.1 | 18 - 19 | 2057 | 67 | |
| XM_015299399.2 | XP_015154885.1 | 19 - 20 | 2201 | 67 | ||
| XM_015299400.2 | XP_015154886.1 | 19 - 20 | 2219 | 67 | ||
| XM_015299401.2 | XP_015154887.1 | 19 - 20 | 2203 | 67 | ||
| XM_015299402.2 | XP_015154888.1 | 19 - 20 | 2189 | 67 | ||
More Information
Set Membership: |
|

EFCAB3
Genomic Map