Assay Details
| Gene Symbol: LOC100859124 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003642876.4 | XP_003642924.3 | 6 - 7 | 2327 | 132 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100859124| Gene Symbol: LOC100859124 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003642876.4 | XP_003642924.3 | 6 - 7 | 2327 | 132 | |
Set Membership: |
|