Assay Details
| Gene Symbol: SHROOM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015278366.2 | XP_015133852.1 | 7 - 8 | 3063 | 66 | |
| XM_015278367.2 | XP_015133853.1 | 7 - 8 | 3021 | 66 | ||
More Information
Set Membership: |
|

SHROOM4
Genomic Map