Assay Details
| Gene Symbol: LOC428714 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015278562.1 | XP_015134048.1 | 7 - 8 | 2491 | 80 | |
| XM_015278563.1 | XP_015134049.1 | 7 - 8 | 2491 | 80 | ||
More Information
Set Membership: |
|

LOC428714
Genomic Map