Assay Details
| Gene Symbol: LOC101748329 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025149217.1 | XP_025004985.1 | 2 - 3 | 2361 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025149217.1 transcripts.
Set Membership: |
|

LOC101748329
Genomic Map